
Methyl 2,6-dimethylisonicotinate
Catalog # APOSOR46524-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:165.189 g/mol
Melting Pt:45…47 °C
:GHS07
Storage Temperature:Ambient
CAS Number:142896-15-9
Formula:C₉H₁₁NO₂
MDL Number:MFCD11656351