Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
Ethyl 2-(chloromethyl)-1,3-thiazole-4-carboxylate 98%
Catalog # APOSOR46233-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl 2-(chloromethyl)-1,3-thiazole-4-carboxylate 98%
Catalog # APOSOR46233-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :205.662 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:842130-48-7
Formula:C₇H₈ClNO₂S
MDL Number:MFCD06761633