
2,4,6-Tris(trichloromethyl)-1,3,5-triazine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6542-67-2
:
Formula:C₆Cl₉N₃
MDL Number:MFCD00023177
:433.161 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together