Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
rel-[(1R,3R)-3-(Aminomethyl)cyclopentyl]carbamic acid 1,1-dimethylethyl ester
Catalog # APOSOR460006-25MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
rel-[(1R,3R)-3-(Aminomethyl)cyclopentyl]carbamic acid 1,1-dimethylethyl ester
Catalog # APOSOR460006-25MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :214.305 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:862700-45-6
Formula:C₁₁H₂₂N₂O₂
MDL Number:MFCD24842666