S-N1,N1-Dimethyl-3-phenylpropane-1,2-diamine dihydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:29802-26-4
:
Formula:C₁₁H₁₈N₂·2ClH
MDL Number:MFCD21607233
:251.196 g/mol
Storage Temperature:Ambient
Frequently Bought Together