
4-Amino-2,4-dihydro-5-(1-methylethyl)-3H-1,2,4-triazol-3-one 95%
Catalog # APOSOR45194-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:142.159 g/mol
Melting Pt:168…172 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:96240-10-7
Formula:C₅H₁₀N₄O
MDL Number:MFCD04972517