
(2S)-2-Methylmorpholine hydrochloride
Catalog # APOSOR45132-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:137.608 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1147108-99-3
Formula:C₅H₁₁NO·ClH
MDL Number:MFCD11506701