Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(R)-3-Benzylmorpholine hydrochloride
(R)-3-Benzylmorpholine hydrochloride
(R)-3-Benzylmorpholine hydrochloride
(R)-3-Benzylmorpholine hydrochloride
APOSOR451244-1G
:
Apollo Scientific
:
(R)-3-Benzylmorpholine hydrochloride
APOSOR451244-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
213.704 g/mol
:
Ambient
:
C₁₁H₁₅NO·ClH
:
MFCD18633581