2-(N-Boc-piperazino)-2-(4-chlorophenyl)ethylamine
Catalog # APOSOR451242-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:339.86 g/mol
Storage Temperature:Ambient
CAS Number:444892-51-7
Formula:C₁₇H₂₆ClN₃O₂
MDL Number:MFCD10015095