4-(Aminomethyl)tetrahydro-2H-pyran-4-carboxylic acid hydrochloride
Catalog # APOSOR451224-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:195.644 g/mol
Storage Temperature:Ambient
CAS Number:1485427-12-0
Formula:C₇H₁₃NO₃·ClH
MDL Number:MFCD25371776