3-(Tetrahydropyran-4-yloxy)benzoic acid
Catalog # APOSOR451222-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:222.237 g/mol
Storage Temperature:Ambient
CAS Number:862088-66-2
Formula:C₁₂H₁₄O₄
MDL Number:MFCD19611449