8-Methylene-1,4-dioxa-spiro[4.5]decane
Catalog # APOSOR451154-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:154.206 g/mol
Storage Temperature:Ambient
CAS Number:51656-90-7
Formula:C₉H₁₄O₂
MDL Number:MFCD07779440