Order Entry
Czech Republic
ContactUsLinkComponent
 
Tetrahydro-2H-thiopyran-4-methanol 1,1-dioxide
Catalog # APOSOR451050-1G
Supplier:  Apollo Scientific
CAS Number:  
Tetrahydro-2H-thiopyran-4-methanol 1,1-dioxide
Catalog # APOSOR451050-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :164.223 g/mol
Storage Temperature:Ambient
CAS Number:473254-28-3
Formula:C₆H₁₂O₃S
MDL Number:MFCD16036707