4-[(Tetrahydro-2H-pyran-4-yl)oxy]aniline hydrochloride
Catalog # APOSOR451048-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:229.703 g/mol
Storage Temperature:Ambient
CAS Number:917483-71-7
Formula:C₁₁H₁₅NO₂·ClH
MDL Number:MFCD28975199