N-Cyclohexyl-tetrahydro-2H-pyran-4-amine
Catalog # APOSOR451045-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:183.291 g/mol
Storage Temperature:Ambient
CAS Number:859521-69-0
Formula:C₁₁H₂₁NO
MDL Number:MFCD08059245