Order Entry
Czech Republic
ContactUsLinkComponent
N-Cyclobutyl-tetrahydro-2H-pyran-4-amine
N-Cyclobutyl-tetrahydro-2H-pyran-4-amine
Catalog # APOSOR451044-1G
Supplier:  Apollo Scientific
CAS Number:  
N-Cyclobutyl-tetrahydro-2H-pyran-4-amine
Catalog # APOSOR451044-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :155.237 g/mol
Storage Temperature:Ambient
CAS Number:885280-95-5
Formula:C₉H₁₇NO
MDL Number:MFCD08059244