Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Boc-3-(1-hydroxyethyl)-azetidine
1-Boc-3-(1-hydroxyethyl)-azetidine
1-Boc-3-(1-hydroxyethyl)-azetidine
APOSOR451035-1G
:
Apollo Scientific
:
1-Boc-3-(1-hydroxyethyl)-azetidine
APOSOR451035-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
201.263 g/mol
:
Ambient
:
1138331-90-4
:
C₁₀H₁₉NO₃
:
MFCD17015912