Ethyl 6-chloro-3-nitroimidazo[1,2-a]pyridine-2-carboxylate
Catalog # APOSOR450072-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:269.641 g/mol
Storage Temperature:Ambient
CAS Number:67625-30-3
Formula:C₁₀H₈ClN₃O₄
MDL Number:MFCD07021399