Order Entry
Czech Republic
ContactUsLinkComponent
2-(Chloromethyl)-3,5-dimethylpyridine hydrochloride 97%
Corrosive
Irritant
2-(Chloromethyl)-3,5-dimethylpyridine hydrochloride 97%
Catalog # APOSOR450036-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
2-(Chloromethyl)-3,5-dimethylpyridine hydrochloride 97%
Catalog # APOSOR450036-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :192.086 g/mol
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:73590-93-9
Formula:C₈H₁₀ClN·ClH
MDL Number:MFCD11112031