0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
[5-(Thien-2-yl)-1H-pyrazol-3-yl]methanol
[5-(Thien-2-yl)-1H-pyrazol-3-yl]methanol
[5-(Thien-2-yl)-1H-pyrazol-3-yl]methanol
APOSOR4423-1G
:
Apollo Scientific
:
[5-(Thien-2-yl)-1H-pyrazol-3-yl]methanol
APOSOR4423-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
180.227 g/mol
:
126…128 °C
:
GHS07
:
Ambient
:
852228-02-5
:
C₈H₈N₂OS
:
MFCD07368555