Order Entry
Czech Republic
ContactUsLinkComponent
6-Bromo-1-cyclopropyl-1,3-dihydro-2H-benzimidazol-2-one
Irritant
6-Bromo-1-cyclopropyl-1,3-dihydro-2H-benzimidazol-2-one
Catalog # APOSOR42260-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-Bromo-1-cyclopropyl-1,3-dihydro-2H-benzimidazol-2-one
Catalog # APOSOR42260-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :253.095 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1416714-45-8
Formula:C₁₀H₉BrN₂O
MDL Number:MFCD22689771