7-Methoxyquinoline-3-carboxylic acid
Catalog # APOSOR42251-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:203.194 g/mol
Storage Temperature:Ambient
CAS Number:474659-26-2
Formula:C₁₁H₉NO₃
MDL Number:MFCD09787873