Order Entry
Czech Republic
ContactUsLinkComponent
2-Bromothiazole-4-carboxamide 97%
Irritant
2-Bromothiazole-4-carboxamide 97%
Catalog # APOSOR42063-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Bromothiazole-4-carboxamide 97%
Catalog # APOSOR42063-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :207.048 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:848501-94-0
Formula:C₄H₃BrN₂OS
MDL Number:MFCD11870082