
(2S)-4-Iodo-2-isopropyl-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2366997-22-8
:
Formula:C₁₀H₁₃IN₂
MDL Number:MFCD31793561
:288.128 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together