Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl 4-chloro-1,3-dimethylpyrazolo[3,4-b]pyridine-5-carboxylate
Ethyl 4-chloro-1,3-dimethylpyrazolo[3,4-b]pyridine-5-carboxylate
Catalog # APOSOR4060-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 4-chloro-1,3-dimethylpyrazolo[3,4-b]pyridine-5-carboxylate
Catalog # APOSOR4060-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :253.685 g/mol
Storage Temperature:Ambient
CAS Number:20481-15-6
Formula:C₁₁H₁₂ClN₃O₂
MDL Number:MFCD00067954