
2-(Chloromethyl)-4,6-dimethoxypyrimidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:114108-86-0
:
Formula:C₇H₉ClN₂O₂
MDL Number:MFCD00274539
:188.612 g/mol
Melting Pt:39…42 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together