

3-(But-1-yl)heptan-2-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:997-69-3
:
Formula:C₁₁H₂₂O
MDL Number:MFCD00027255
:170.292 g/mol
Boiling Pt:85…88 °C (8 mm Hg)
Density:0.82 g/cm³ (25 °C)
Storage Temperature:Refrigerator
UN:1993
ADR:3,III
:GHS02
:GHS07
Frequently Bought Together