Order Entry
Czech Republic
ContactUsLinkComponent
2-(Bromomethyl)-4,6-dimethylpyridine 95%
Irritant
2-(Bromomethyl)-4,6-dimethylpyridine 95%
Catalog # APOSOR40233-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(Bromomethyl)-4,6-dimethylpyridine 95%
Catalog # APOSOR40233-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :200.076 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:79313-01-2
Formula:C₈H₁₀BrN
MDL Number:MFCD10699165