
Methyl 4-bromo-2-iodobenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1093418-75-7
:
Formula:C₈H₆BrIO₂
MDL Number:MFCD16877107
:340.941 g/mol
Melting Pt:50 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together