N-Methoxy-N,1-dimethylpiperidine-4-carboxamide 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:215950-19-9
:
Formula:C₉H₁₈N₂O₂
MDL Number:MFCD22575146
:186.251 g/mol
Storage Temperature:Ambient
Frequently Bought Together