
Methyl 2-bromo-5-formylbenzoate
Catalog # APOSOR400745-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:243.054 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1043418-95-6
Formula:C₉H₇BrO₃
MDL Number:MFCD26863463