Order Entry
Czech Republic
ContactUsLinkComponent
N-[2-(Diethylamino)ethyl]-2,4-dimethyl-5-formyl-1H-pyrrole-3-carboxamide 98%
Irritant
N-[2-(Diethylamino)ethyl]-2,4-dimethyl-5-formyl-1H-pyrrole-3-carboxamide 98%
Catalog # APOSOR40055-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
N-[2-(Diethylamino)ethyl]-2,4-dimethyl-5-formyl-1H-pyrrole-3-carboxamide 98%
Catalog # APOSOR40055-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :265.351 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:356068-86-5
Formula:C₁₄H₂₃N₃O₂
MDL Number:MFCD12913910