3-Bromo-5-chlorobenzene -1,2-diamine 98%
Catalog # APOSOR400386-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:221.482 g/mol
Storage Temperature:Ambient
CAS Number:500862-39-5
Formula:C₆H₆BrClN₂
MDL Number:MFCD15144647