
4-(Bromomethyl)-2-chlorobenzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:290820-38-1
:
Formula:C₈H₆BrClO₂
MDL Number:MFCD18378710
:249.489 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together