Methyl 3-bromo-5-methylbenzoate
Catalog # APOSOR400190-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:229.071 g/mol
Storage Temperature:Ambient
CAS Number:478375-40-5
Formula:C₉H₉BrO₂
MDL Number:MFCD11007827