Order Entry
Czech Republic
ContactUsLinkComponent
2-Iodo-6-methoxybenzoic acid 97%
2-Iodo-6-methoxybenzoic acid 97%
Catalog # APOSOR400154-250MG
Supplier:  Apollo Scientific
CAS Number:  
2-Iodo-6-methoxybenzoic acid 97%
Catalog # APOSOR400154-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :278.044 g/mol
Storage Temperature:Ambient
CAS Number:66195-39-9
Formula:C₈H₇IO₃
MDL Number:MFCD13562947