
Methyl 2-bromo-3-methoxybenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:59453-47-3
:
Formula:C₉H₉BrO₃
MDL Number:MFCD14697991
:245.07 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together