3,5-Dinitro-4-methoxybenzenesulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175203-74-4
:
Formula:C₇H₅ClN₂O₇S
MDL Number:MFCD00178761
:296.642 g/mol
Storage Temperature:Refrigerator
UN:2928
ADR:6.1+8,I
Frequently Bought Together