1-(1-Methylcyclopropyl)ethanamine hydrochloride ≥98%
Catalog # APOSOR370099-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:135.635 g/mol
Storage Temperature:Ambient
CAS Number:54343-93-0
Formula:C₆H₁₃N·ClH
MDL Number:MFCD09971518