Order Entry
Czech Republic
ContactUsLinkComponent
3-Methyl-4-(piperidin-4-yl)aniline ≥98%
3-Methyl-4-(piperidin-4-yl)aniline ≥98%
Catalog # APOSOR370079-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Methyl-4-(piperidin-4-yl)aniline ≥98%
Catalog # APOSOR370079-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :190.285 g/mol
Storage Temperature:Ambient
CAS Number:1260864-79-6
Formula:C₁₂H₁₈N₂
MDL Number:MFCD17486190