N-Propyl-4,5,6,7-tetrahydrobenzo[d]thiazol-6-amine hydrochloride ≥98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1415559-98-6
:
Formula:C₁₀H₁₆N₂S·ClH
MDL Number:MFCD22572793
:232.773 g/mol
Storage Temperature:Ambient
Frequently Bought Together