Order Entry
Czech Republic
ContactUsLinkComponent
N,3-Dimethylbutan-2-amine ≥98%
N,3-Dimethylbutan-2-amine ≥98%
Catalog # APOSOR361732-1G
Supplier:  Apollo Scientific
CAS Number:  
N,3-Dimethylbutan-2-amine ≥98%
Catalog # APOSOR361732-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :101.19 g/mol
Storage Temperature:Ambient
CAS Number:34317-39-0
Formula:C₆H₁₅N
MDL Number:MFCD09047478