
4-Carboxy-3-methylphenylboronic acid, pinacol ester 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:890839-22-2
:
Formula:C₁₄H₁₉BO₄
MDL Number:MFCD09037836
:262.109 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together