Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(1H-Indol-3-yl)-2-methylpropanoic acid
2-(1H-Indol-3-yl)-2-methylpropanoic acid 98%
2-(1H-Indol-3-yl)-2-methylpropanoic acid 98%
APOSOR350487-250MG
:
Apollo Scientific
:
2-(1H-Indol-3-yl)-2-methylpropanoic acid 98%
APOSOR350487-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
203.237 g/mol
:
Ambient
:
2770-92-5
:
C₁₂H₁₃NO₂
:
MFCD17011803