Order Entry
Czech Republic
ContactUsLinkComponent
(S)-N6-Propyl-4,5,6,7-tetrahydrobenzothiazole-2,6-diamine 98%
(S)-N6-Propyl-4,5,6,7-tetrahydrobenzothiazole-2,6-diamine 98%
Catalog # APOSOR350364-1G
Supplier:  Apollo Scientific
CAS Number:  
(S)-N6-Propyl-4,5,6,7-tetrahydrobenzothiazole-2,6-diamine 98%
Catalog # APOSOR350364-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :211.327 g/mol
Storage Temperature:Ambient
CAS Number:104632-26-0
Formula:C₁₀H₁₇N₃S
MDL Number:MFCD00869076