9,10-Bis(4-methoxyphenyl)-2-chloroanthracene
Catalog # APOSOR350066-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:424.918 g/mol
Storage Temperature:Ambient
CAS Number:110904-87-5
Formula:C₂₈H₂₁ClO₂
MDL Number:MFCD00009935