Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Imidazo[1,2-a]pyridine-5-carboxaldehyde
Imidazo[1,2-a]pyridine-5-carboxaldehyde 95+%
Imidazo[1,2-a]pyridine-5-carboxaldehyde 95+%
Imidazo[1,2-a]pyridine-5-carboxaldehyde 95+%
APOSOR346647-1G
:
Apollo Scientific
:
Imidazo[1,2-a]pyridine-5-carboxaldehyde 95+%
APOSOR346647-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
146.146 g/mol
:
Ambient
:
372147-50-7
:
C₈H₆N₂O
:
MFCD10698080