Order Entry
Czech Republic
ContactUsLinkComponent
2-Bromo-1-(3-methoxyphenyl)propan-1-one 95+%
2-Bromo-1-(3-methoxyphenyl)propan-1-one 95+%
Catalog # APOSOR346564-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-1-(3-methoxyphenyl)propan-1-one 95+%
Catalog # APOSOR346564-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :243.097 g/mol
Storage Temperature:Ambient
CAS Number:21726-71-6
Formula:C₁₀H₁₁BrO₂
MDL Number:MFCD12031564