1-(2-Methoxyphenyl)propan-1-one 95+%
Catalog # APOSOR346562-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:164.201 g/mol
Storage Temperature:Ambient
CAS Number:5561-92-2
Formula:C₁₀H₁₂O₂
MDL Number:MFCD00048613