Order Entry
Czech Republic
ContactUsLinkComponent
3-Bromo-5-(1,3-dioxolan-2-yl)phenylamine 95+%
3-Bromo-5-(1,3-dioxolan-2-yl)phenylamine 95+%
Catalog # APOSOR346555-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Bromo-5-(1,3-dioxolan-2-yl)phenylamine 95+%
Catalog # APOSOR346555-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :244.085 g/mol
Storage Temperature:Ambient
CAS Number:936844-19-8
Formula:C₉H₁₀BrNO₂
MDL Number:MFCD29918682